1,1,3,3-Tetramethyl-2-thiourea 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N,N',N'-Tetramethylthiourea, Tetramethyl-2-thiourea, Tetramethylthiourea
L13392-06
L13392-14
CAAAAL13392-06EA
41.71
CAD
CAAAAL13392-06
CAAAAL13392-14
1,1,3,3-Tetramethyl-2-thiourea 98%
1,1,3,3-Tetramethyl-2-thiourea
Purity: 98%
CAS No.: 2782-91-4
Molecular Formula: C5H12N2S
Formula Weight: 132.23
Flash Point: 190°(374°F)
Odor: Odorless
Boiling Point: 245°
Beilstein Reference: 1744916
MDL No.: MFCD00008324
Formula:
C₅H₁₂N₂S MW: 132.23 g/mol Boiling Pt: 245 °C Melting Pt: 75…77 °C Flash Pt: 190 °C(374 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008324 CAS Number: 2782-91-4 EINECS: 220-488-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...