Dodecyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13380-03
L13380-06
CAAAAL13380-03EA
52.61
CAD
CAAAAL13380-03
CAAAAL13380-06
Dodecyl isocyanate 98%
Dodecyl isocyanate
Purity: 98%
CAS No.: 4202-38-4
Molecular Formula: CH3(CH2)11NCO
Formula Weight: 211.35
Storage Temperature: 2°C to 8°C
Flash Point: 134°(273°F)
Refractive Index: 1.4410
Odor: Irritating
Boiling Point: 157-161°/21mm
Beilstein Reference: 1774823
UN No.: 2206
MDL No.: MFCD00041744
Formula:
C₁₃H₂₅NO Boiling Pt: 157…161 °C/21 mm Density: 0.877 Flash Pt: 134 °C (273 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00041744 CAS Number: 4202-38-4 EINECS: 224-111-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...