Trimethyl orthoacetate 98%

Supplier: Thermo Scientific Chemicals
Danger

L13369-AD L13369-AK
CAAAAL13369-ADEA 73 CAD
CAAAAL13369-AD CAAAAL13369-AK
Trimethyl orthoacetate 98%
Trimethyl orthoacetate

Purity: 98% CAS No.: 1445-45-0 Molecular Formula: C5H12O3 Formula Weight: 120.15 Flash Point: 16°(60°F) Refractive Index: 1.3890 Boiling Point: 107-109° Beilstein Reference: 1098338 UN No.: 3272 MDL No.: MFCD00008477

Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 107…109 °C
Melting Pt: –58 °C
Density: 0.956
Flash Pt: 16 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008477
CAS Number: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR