1-Boc-piperazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13363-03
L13363-06
L13363-14
CAAAAL13363-03EA
55.62
CAD
CAAAAL13363-03
CAAAAL13363-06
CAAAAL13363-14
1-Boc-piperazine 99%
1-Boc-piperazine
Purity: 99%
CAS No.: 57260-71-6
Molecular Formula: C9H18N2O2
Formula Weight: 186.26
Melting Point: 44-48°
Beilstein Reference: 879985
Formula:
C₉H₁₈N₂O₂ Melting Pt: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075265 CAS Number: 57260-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...