6-Nitro-m-anisic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L13346-06 L13346-14
CAAAAL13346-06EA 35.71 CAD
CAAAAL13346-06 CAAAAL13346-14
6-Nitro-m-anisic acid 97%
6-Nitro-m-anisic acid

Purity: 97% CAS No.: 1882-69-5 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 130-135° Beilstein Reference: 1877657

Formula: C₈H₇NO₅
Melting Pt: 130…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00151836
CAS Number: 1882-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR