(4-Iodophenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13345-03
L13345-06
CAAAAL13345-03EA
64.46
CAD
CAAAAL13345-03
CAAAAL13345-06
(4-Iodophenyl)acetic acid 97%
(4-Iodophenyl)acetic acid
Purity: 97%
CAS No.: 1798-06-7
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Melting Point: 136-140°
Beilstein Reference: 1940558
Formula:
(C₆H₄I)CH₂COOH Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082985 CAS Number: 1798-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...