2-Bromo-4'-(diethylamino)acetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13337-03
L13337-06
CAAAAL13337-03EA
109.97
CAD
CAAAAL13337-03
CAAAAL13337-06
2-Bromo-4'-(diethylamino)acetophenone 98%
Purity: 98%
CAS No.: 207986-25-2
Molecular Formula: C12H16BrNO
Formula Weight: 270.18
Melting Point: 50-52°
Beilstein Reference: 7921481
Formula:
C₁₂H₁₆BrNO Melting Pt: 50…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151798 CAS Number: 207986-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...