Xylene (mixture of isomers) 97+%

Supplier: Thermo Scientific Chemicals
Danger

L13317-0F L13317-AP L13317-AU
CAAAAL13317-0FEA 106.81 CAD
CAAAAL13317-0F CAAAAL13317-AP CAAAAL13317-AU
Xylene (mixture of isomers) 97+%
Xylene (mixture of isomers)

Purity: 97+% CAS No.: 1330-20-7 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 29°(84°F) Refractive Index: 1.4970 Odor: Aromatic Melting Point: -25° Boiling Point: 137-144° Beilstein Reference: 1901563 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00077264

Formula: C₆H₄(CH₃)₂
Boiling Pt: 137…144 °C
Melting Pt: < –25 °C
Density: 0.860
Flash Pt: 29 °C(84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 14,10081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR