4-Vinylpyridine 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ) 80 -150 ppm
L13316-AC
L13316-AE
L13316-AP
CAAAAL13316-ACEA
54.67
CAD
CAAAAL13316-AC
CAAAAL13316-AE
CAAAAL13316-AP
4-Vinylpyridine 95% stabilized
4-Vinylpyridine
Purity: 95%
CAS No.: 100-43-6
Molecular Formula: C7H7N
Formula Weight: 105.14
Storage Temperature: -30°C to -10°C
Flash Point: 51°(124°F)
Refractive Index: 1.5510
Boiling Point: 58-61°/12mm
Beilstein Reference: 104506
UN No.: 3073
MDL No.: MFCD00006447
Formula:
C₇H₇N MW: 105.14 g/mol Boiling Pt: 58…61 °C/12 mm Melting Pt: –39 °C Density: 0.984 Flash Pt: 51 °C (124 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00006447 CAS Number: 100-43-6 EINECS: 202-852-0 UN: 3073 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...