Polysorbate 80

Supplier: Thermo Scientific Chemicals

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

L13315-0F L13315-AE L13315-AP
CAAAAL13315-0FEA 259.12 CAD
CAAAAL13315-0F CAAAAL13315-AE CAAAAL13315-AP
Polysorbate 80
Polysorbate 80

Used for solubilizing membrane proteins during isolation and purification


CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107

Boiling Pt: >100 °C
Melting Pt: 100 °C
Density: 1.064
Flash Pt: >149 °C(300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 14,7582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR