Sodium peroxide 95%

Supplier: Thermo Scientific Chemicals
Danger

L13306-0B L13306-18 L13306-30
CAAAAL13306-0BEA 824.76 CAD
CAAAAL13306-0B CAAAAL13306-18 CAAAAL13306-30
Sodium peroxide 95%
Sodium peroxide

Purity: 95% CAS No.: 1313-60-6 Molecular Formula: Na2O2 Formula Weight: 77.98 Melting Point: 460° dec. Merck Reference: 14,8655 MDL No.: MFCD00003497

Formula: Na₂O₂
Melting Pt: 460 °C (dec.)
Density: 2.805
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 14,8655

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR