Tetrabutylammonium fluoride trihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

L13303-09 L13303-18
CAAAAL13303-09EA 111.07 CAD
CAAAAL13303-09 CAAAAL13303-18
Tetrabutylammonium fluoride trihydrate 98%
Tetrabutylammonium fluoride trihydrate

Purity: 98% CAS No.: 87749-50-6 Molecular Formula: C16H36FN.3H2O Formula Weight: 315.52 (261.47anhy) Odor: Odorless Melting Point: 54-60° Beilstein Reference: 3761900 UN No.: 2811 MDL No.: MFCD00149981

Formula: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Melting Pt: 54…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR