2-Pentanone 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13262-14
L13262-22
L13262-36
CAAAAL13262-14EA
42.66
CAD
CAAAAL13262-14
CAAAAL13262-22
CAAAAL13262-36
2-Pentanone 99+%
2-Pentanone
Purity: 99+%
CAS No.: 107-87-9
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 7°(44°F)
Refractive Index: 1.3905
Melting Point: -78°
Boiling Point: 100-101°
Beilstein Reference: 506058
Merck Reference: 14,6114
UN No.: 1249
MDL No.: MFCD00009400
Formula:
C₅H₁₀O Boiling Pt: 100…101 °C Melting Pt: –78 °C Density: 0.813 Flash Pt: 7 °C (44 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009400 CAS Number: 107-87-9 EINECS: 203-528-1 UN: 1249 ADR: 3,II Merck Index: 14,6114 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...