N-Octyl-β-D-glucopyranoside
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13259-03
L13259-MD
CAAAAL13259-03EA
167.48
CAD
CAAAAL13259-03
CAAAAL13259-MD
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Non-ionic detergent for membrane proteins
CAS No.: 29836-26-8
Molecular Formula: C14H28O6
Formula Weight: 292.37
Melting Point: 103-104°
Beilstein Reference: 84118
Optical Rotation: -31° (c=2 in methanol)
Formula:
C₁₄H₂₈O₆ Melting Pt: 103…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063288 CAS Number: 29836-26-8 EINECS: 249-887-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...