Fluorescein 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid

L13251-22 L13251-36
CAAAAL13251-22EA 70.31 CAD
CAAAAL13251-22 CAAAAL13251-36
Fluorescein 90+%
Fluorescein

Purity: 90+% CAS No.: 2321-07-5 Molecular Formula: C20H12O5 Formula Weight: 332.32 Melting Point: 320° Beilstein Reference: 94324 Merck Reference: 14,4159 MDL No.: MFCD00005050

Formula: C₂₀H₁₂O₅
MW: 332.31 g/mol
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00005050
CAS Number: 2321-07-5
EINECS: 219-031-8
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b9d8d37f0106f4c8ef2d7759759d21bd43cc03&vid=25b78df95a04ab26c5ba8c96b95f031056926b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR