(S)-(-)-Limonene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
Purity: 97%
CAS No.: 5989-54-8
Molecular Formula: C10H16
Formula Weight: 136.23
Flash Point: 48°(118°F)
Refractive Index: 1.4720
Melting Point: -74°
Boiling Point: 175-177°
Beilstein Reference: 2323991
Merck Reference: 14,5493
Optical Rotation: -90° (c=10 in ethanol)
UN No.: 2052
MDL No.: MFCD00001558
Formula:
C₁₀H₁₆ Boiling Pt: 175…177 °C Melting Pt: –74 °C Density: 0.844 Flash Pt: 48 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001558 CAS Number: 5989-54-8 EINECS: 227-815-6 UN: 2052 ADR: 3,III Merck Index: 14,5493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...