D-(-)-Leucinol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

L13243-03 L13243-06
CAAAAL13243-03EA 77.42 CAD
CAAAAL13243-03 CAAAAL13243-06
D-(-)-Leucinol 97%
D(-)-Leucinol

Purity: 97% CAS No.: 53448-09-2 Molecular Formula: C6H15NO Formula Weight: 117.19 Flash Point: 90°(194°F) Refractive Index: 1.4496 Boiling Point: 198-200°/690mm Beilstein Reference: 1719241 Optical Rotation: -3.2° (c=9 in ethanol) MDL No.: MFCD00004734

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199
Density: 0.917
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004734
CAS Number: 53448-09-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR