L(+)-Lactic acid, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13242-06
L13242-14
L13242-22
CAAAAL13242-06EA
121.98
CAD
CAAAAL13242-06
CAAAAL13242-14
CAAAAL13242-22
L(+)-Lactic acid, anhydrous 98%
L(+)-Lactic acid
Useful chiral synthon, building block for depsipeptides
Purity: 98%
CAS No.: 79-33-4
Molecular Formula: C3H6O3
Formula Weight: 90.08
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₃H₆O₃ Boiling Pt: 119 °C/12 mm Melting Pt: 52…54 °C Density: 1.22 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064266 CAS Number: 79-33-4 EINECS: 201-196-2 Merck Index: 14,5337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...