Isobutyric anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylpropanoic anhydride, 2-Methylpropionic anhydride
L13240-0F
L13240-AE
L13240-AP
CAAAAL13240-0FEA
240.16
CAD
CAAAAL13240-0F
CAAAAL13240-AE
CAAAAL13240-AP
Isobutyric anhydride 97%
Isobutyric anhydride
Purity: 97%
CAS No.: 97-72-3
Molecular Formula: [(CH3)2CHCO]2O
Formula Weight: 158.20
Flash Point: 67°(152°F)
Refractive Index: 1.4060
Odor: Pungent
Melting Point: -56°
Boiling Point: 182°
Beilstein Reference: 386267
UN No.: 2922
MDL No.: MFCD00008913
Formula:
C₈H₁₄O₃ MW: 158.2 g/mol Boiling Pt: 182 °C Melting Pt: -69 Density: 0.954 Flash Pt: 67 °C (152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008913 CAS Number: 97-72-3 EINECS: 202-603-6 UN: 2922 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...