Hexane (mixture of isomers) 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexanes, Isohexane

L13233-0F L13233-AP L13233-AU
CAAAAL13233-0FEA 120.4 CAD
CAAAAL13233-0F CAAAAL13233-AP CAAAAL13233-AU
Hexane (mixture of isomers) 98+%
Hexane (mixture of isomers)

Purity: 98+% CAS No.: 92112-69-1 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Refractive Index: 1.3795 Melting Point: -95° Boiling Point: 65-70° UN No.: 1208 MDL No.: MFCD02179311

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 65…70 °C
Melting Pt: –95 °C
Density: 0.67
Flash Pt: <–23 °C (–9 °F)
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 295-570-2
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR