Calcium chloride, dried 97%, powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

L13191-0B L13191-0I L13191-30
CAAAAL13191-0BEA 121.34 CAD
CAAAAL13191-0B CAAAAL13191-0I CAAAAL13191-30
Calcium chloride, dried 97%, powder
Calcium chloride

Purity: 97% CAS No.: 10043-52-4 Molecular Formula: CaCl2 Formula Weight: 110.99 Melting Point: 782° Boiling Point: 1600° Merck Reference: 14,1659 MDL No.: MFCD00010903

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: >1600 °C
Melting Pt: 782 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,1659

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR