Butyric acid 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13189-0F
L13189-AE
L13189-AP
CAAAAL13189-0FEA
124.98
CAD
CAAAAL13189-0F
CAAAAL13189-AE
CAAAAL13189-AP
Butyric acid 99+%
Butyric acid
Purity: 99+%
CAS No.: 107-92-6
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: 69°(156°F)
Refractive Index: 1.3980
Odor: Rancid
pH: 2.5
Melting Point: -8°
Boiling Point: 162-164°
Beilstein Reference: 906770
Merck Reference: 14,1593
UN No.: 2820
MDL No.: MFCD00002814
Formula:
C₄H₈O₂ Boiling Pt: 162…164 °C Melting Pt: –8 °C Density: 0.964 Flash Pt: 69 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002814 CAS Number: 107-92-6 EINECS: 203-532-3 UN: 2820 ADR: 8,III Merck Index: 14,1593 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...