Methyl ethyl ketone 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13185-0F
L13185-AP
L13185-AU
CAAAAL13185-0FEA
140.15
CAD
CAAAAL13185-0F
CAAAAL13185-AP
CAAAAL13185-AU
Methyl ethyl ketone 99%
Methyl ethyl ketone
Purity: 99%
CAS No.: 78-93-3
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -3°(26°F)
Refractive Index: 1.3785
Odor: Characteristic
Melting Point: -87°
Boiling Point: 80°
Beilstein Reference: 741880
Merck Reference: 14,6072
UN No.: 1193
MDL No.: MFCD00011648
Formula:
C₂H₅COCH₃ Boiling Pt: 80 °C Melting Pt: –87 °C Density: 0.805 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II REACH: 01-2119457290-43 Merck Index: 14,6072 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...