Barium manganate tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13172-06
L13172-14
CAAAAL13172-06EA
50.24
CAD
CAAAAL13172-06
CAAAAL13172-14
Barium manganate tech. 90%, Technical Grade
Barium manganate
Purity: 90%
CAS No.: 7787-35-1
Molecular Formula: BaMnO4
Formula Weight: 256.28
Merck Reference: 14,981
UN No.: 3087
MDL No.: MFCD00011514
Formula:
BaMnO₄ Density: 4.850 Storage Temperature: Ambient |
MDL Number:
MFCD00011514 CAS Number: 7787-35-1 EINECS: 232-109-6 UN: 3087 ADR: 5.1,III Merck Index: 14,981 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...