1-Butanol 99%

Supplier: Thermo Scientific Chemicals
Danger

L13171-0F L13171-AE L13171-AU
CAAAAL13171-0FEA 126.56 CAD
CAAAAL13171-0F CAAAAL13171-AE CAAAAL13171-AU
1-Butanol 99%
1-Butanol

Purity: 99% CAS No.: 71-36-3 Molecular Formula: CH3(CH2)3OH Formula Weight: 74.12 Flash Point: 35°(95°F) Refractive Index: 1.3990 Odor: Alcohol-like pH: 7 Melting Point: -90 to -89° Boiling Point: 117-118° Beilstein Reference: 969148 Merck Reference: 14,1540 UN No.: 1120 MDL No.: MFCD00002964

Formula: H₃C(CH₂)₂CH₂OH
Boiling Pt: 117…118 °C
Melting Pt: –90 to –89 °C
Density: 0.810
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 14,1540
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR