Ammonia solution 28%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

L13168-0F L13168-AP L13168-AU
CAAAAL13168-0FEA 114.39 CAD
CAAAAL13168-0F CAAAAL13168-AP CAAAAL13168-AU
Ammonia solution 28%
Ammonia solution

CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Odor: Ammonia-like pH: 12 Merck Reference: 14,494 MDL No.: MFCD00011418

Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Density: 0.90
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III
Merck Index: 14,494

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR