6-Methoxysalicylaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L13145-03
L13145-06
CAAAAL13145-03EA
122.45
CAD
CAAAAL13145-03
CAAAAL13145-06
6-Methoxysalicylaldehyde 98+%
6-Methoxysalicylaldehyde
Purity: 98+%
CAS No.: 700-44-7
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 68-74°
Beilstein Reference: 1101524
Formula:
C₈H₈O₃ Melting Pt: 68…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151830 CAS Number: 700-44-7 EINECS: 211-844-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...