4-Anisoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methoxybenzoyl chloride

L13120-14 L13120-22
CAAAAL13120-14EA 75.05 CAD
CAAAAL13120-14 CAAAAL13120-22
4-Anisoyl chloride 97%
4-Anisoyl chloride

Purity: 97% CAS No.: 100-07-2 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Flash Point: 87°(188°F) Refractive Index: 1.5810 Melting Point: 22° Boiling Point: 145°/14mm Beilstein Reference: 471918 Merck Reference: 14,673 UN No.: 1729 MDL No.: MFCD00000687

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 145 °C/14 mm
Melting Pt: 22 °C
Density: 1.260
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000687
CAS Number: 100-07-2
EINECS: 202-816-4
UN: 1729
ADR: 8,II
Merck Index: 14,673

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR