4-Nitro-m-toluidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methyl-4-nitroaniline, 4-Nitro-3-methylaniline, 5-Amino-2-nitrotoluene

L13112-06 L13112-14
CAAAAL13112-06EA 124.35 CAD
CAAAAL13112-06 CAAAAL13112-14
4-Nitro-m-toluidine 99%
4-Nitro-m-toluidine

Purity: 99% CAS No.: 611-05-2 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Odor: Characteristic Melting Point: 133-137° Beilstein Reference: 2209112 UN No.: 2660 MDL No.: MFCD00091833

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00091833
CAS Number: 611-05-2
EINECS: 210-247-8
UN: 2660
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR