5,5-Dimethylacetylacetone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5,5-Dimethylhexane-2,4-dione, 2,2-Dimethyl-3,5-hexanedione, 5,5-Diméthylhexanedione-2,4
L13104-03
L13104-06
CAAAAL13104-03EA
140.94
CAD
CAAAAL13104-03
CAAAAL13104-06
5,5-Dimethylacetylacetone 99%
5,5-Dimethylacetylacetone
Purity: 99%
CAS No.: 7307-04-2
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 57°(134°F)
Refractive Index: 1.4600
Boiling Point: 60°/10mm
Beilstein Reference: 969813
UN No.: 1224
MDL No.: MFCD00142767
Formula:
C₈H₁₄O₂ MW: 142.2 g/mol Boiling Pt: 60 °C (10 mmHg) Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00142767 CAS Number: 7307-04-2 EINECS: 230-760-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...