Hydrochloric acid 36% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

L13091-0F L13091-AU
CAAAAL13091-0FEA 101.44 CAD
CAAAAL13091-0F CAAAAL13091-AU
Hydrochloric acid 36% (w/w) in aqueous solution
Hydrochloric acid

CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Odor: Pungent Boiling Point: 81.5-110° Merck Reference: 14,4780 MDL No.: MFCD00011324

Formula: HCl
Boiling Pt: 81.5…110 °C
Melting Pt: –30 °C
Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Merck Index: 14,4780

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR