3-Cyanophenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13045-03
L13045-06
CAAAAL13045-03EA
72.68
CAD
CAAAAL13045-03
CAAAAL13045-06
3-Cyanophenyl isothiocyanate 97%
3-Cyanophenyl isothiocyanate
Purity: 97%
CAS No.: 3125-78-8
Molecular Formula: C8H4N2S
Formula Weight: 160.20
Odor: Irritating
Melting Point: 64-66°
Beilstein Reference: 2803099
UN No.: 2923
MDL No.: MFCD00041070
Formula:
C₈H₄N₂S Boiling Pt: 294 °C (1013 hPa) Melting Pt: 64…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041070 CAS Number: 3125-78-8 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...