3-Penten-2-one tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13031-03
L13031-06
CAAAAL13031-03EA
65.1
CAD
CAAAAL13031-03
CAAAAL13031-06
3-Penten-2-one tech. 85%, Technical Grade
3-Penten-2-one
Purity: 85%
CAS No.: 625-33-2
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 21°(69°F)
Refractive Index: 1.4355
Odor: Pungent, makes eyes water
Boiling Point: 119-121°
Beilstein Reference: 1633505
UN No.: 1992
MDL No.: MFCD00009290
Formula:
C₅H₈O Boiling Pt: 119…121 °C Density: 0.862 Flash Pt: 21 °C(69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009290 CAS Number: 625-33-2 EINECS: 210-888-3 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...