2-Chloro-4-fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

L12957-03 L12957-06
CAAAAL12957-03EA 133.98 CAD
CAAAAL12957-03 CAAAAL12957-06
2-Chloro-4-fluorobenzylamine 97%
2-Chloro-4-fluorobenzylamine

Purity: 97% CAS No.: 15205-11-5 Molecular Formula: C7H7ClFN Formula Weight: 159.59 Flash Point: 110°(230°F) Refractive Index: 1.5350 Boiling Point: 94-95°/15mm Beilstein Reference: 2936421 UN No.: 2735 MDL No.: MFCD00042532

Formula: C₇H₇ClFN
MW: 159.59 g/mol
Boiling Pt: 94…95 °C/15 mm
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042532
CAS Number: 15205-11-5
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR