3-Hydroxy-2-nitrobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12841-04
L12841-09
CAAAAL12841-04EA
202.24
CAD
CAAAAL12841-04
CAAAAL12841-09
CAAAAL12841-ME
3-Hydroxy-2-nitrobenzoic acid 98+%
3-Hydroxy-2-nitrobenzoic acid
Purity: 98+%
CAS No.: 602-00-6
Molecular Formula: C7H5NO5
Formula Weight: 183.12
Melting Point: 177-181°
Beilstein Reference: 2372248
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Melting Pt: 177…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017003 CAS Number: 602-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...