Dibutyl sulfate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di-n-butylsulfate, Di-n-butylsulphate

L12813-09 L12813-18
CAAAAL12813-09EA 51.19 CAD
CAAAAL12813-09 CAAAAL12813-18
Dibutyl sulfate 95%
Dibutyl sulphate

Purity: 95% CAS No.: 625-22-9 Molecular Formula: C8H18O4S Formula Weight: 210.29 Refractive Index: 1.4205 Boiling Point: 115-116°/6mm MDL No.: MFCD00059420

Formula: C₈H₁₈O₄S
MW: 210.29 g/mol
Boiling Pt: 115…116 °C/6 mm
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00059420
CAS Number: 625-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR