2,6-Dimethylphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12805-06
L12805-14
CAAAAL12805-06EA
121.19
CAD
CAAAAL12805-06
CAAAAL12805-14
2,6-Dimethylphenyl isocyanate 98%
2,6-Dimethylphenyl isocyanate
Purity: 98%
CAS No.: 28556-81-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.5356
Boiling Point: 71-72°/3mm
Beilstein Reference: 742593
UN No.: 2206
MDL No.: MFCD00002012
Formula:
C₉H₉NO Boiling Pt: 71…72 °C/3 mm Density: 1.057 Flash Pt: 86 °C (186 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002012 CAS Number: 28556-81-2 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...