(2-Iodophenyl)acetonitrile 96%

Supplier: Thermo Scientific Chemicals
Warning

L12767-03 L12767-06
CAAAAL12767-03EA 48.35 CAD
CAAAAL12767-03 CAAAAL12767-06
(2-Iodophenyl)acetonitrile 96%
(2-Iodophenyl)acetonitrile

Purity: 96% CAS No.: 40400-15-5 Molecular Formula: C8H6IN Formula Weight: 243.05 Flash Point: 110°(230°F) Refractive Index: 1.6170 Boiling Point: 139-140°/13mm Beilstein Reference: 2083106 UN No.: 3276 MDL No.: MFCD00040888

Formula: C₈H₆IN
Boiling Pt: 139…140 °C/13 mm
Density: 1.750
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040888
CAS Number: 40400-15-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR