N,N-Dibutylformamide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formdibutylamide, Formic acid dibutylamide

L12755-14 L12755-22 L12755-36
CAAAAL12755-14EA 56.56 CAD
CAAAAL12755-14 CAAAAL12755-22 CAAAAL12755-36
N,N-Dibutylformamide 99%
N,N-Dibutylformamide

Purity: 99% CAS No.: 761-65-9 Molecular Formula: C9H19NO Formula Weight: 157.26 Flash Point: 115°(239°F) Refractive Index: 1.4445 Melting Point: -30° Boiling Point: 119-120°/15mm Beilstein Reference: 636100 MDL No.: MFCD00003289

Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 119…120 °C/15 mm
Melting Pt: < –30 °C
Density: 0.864
Flash Pt: 115 °C(239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003289
CAS Number: 761-65-9
EINECS: 212-090-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR