(S)-(+)-2-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L12754-03 L12754-06
CAAAAL12754-03EA 130.19 CAD
CAAAAL12754-03 CAAAAL12754-06
(S)-(+)-2-Phenylbutyric acid 99%
(S)-(+)-2-Phenylbutyric acid

Purity: 99% CAS No.: 4286-15-1 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5150 Boiling Point: 270-272° Beilstein Reference: 2045208 Optical Rotation: +100° (c=5 in toluene) MDL No.: MFCD00063164

Formula: C₁₀H₁₂O₂
Boiling Pt: 270…272 °C
Density: 1,055
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063164
CAS Number: 4286-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR