2,6-Toluenediisocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-TDI, 2-Methyl-m-phenylene diisocyanate, Tolylene 2,6-diisocyanate

L12745-03 L12745-06
CAAAAL12745-03EA 101.59 CAD
CAAAAL12745-03 CAAAAL12745-06
2,6-Toluenediisocyanate 97%
2,6-Toluenediisocyanate

Purity: 97% CAS No.: 91-08-7 Molecular Formula: C9H6N2O2 Formula Weight: 174.16 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5708 Odor: Pungent Melting Point: 13° Boiling Point: 129-133°/18mm Beilstein Reference: 2211546 UN No.: 2078 MDL No.: MFCD00002010

Formula: C₉H₆N₂O₂
Boiling Pt: 129…133 °C/18 mm
Melting Pt: 13 °C
Density: 1.226
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002010
CAS Number: 91-08-7
EINECS: 202-039-0
UN: 2078
ADR: "6.1",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR