4-Ethylphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12718-03
L12718-06
L12718-14
CAAAAL12718-03EA
40.13
CAD
CAAAAL12718-03
CAAAAL12718-06
CAAAAL12718-14
4-Ethylphenyl isocyanate 98%
4-Ethylphenyl isocyanate
Purity: 98%
CAS No.: 23138-50-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.5270
Odor: Pungent
Boiling Point: 67-68°/1mm
Beilstein Reference: 971100
UN No.: 2206
MDL No.: MFCD00013881
Formula:
C₉H₉NO Boiling Pt: 67…68 °C/1 mm Density: 1.024 Flash Pt: 78 °C (172 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013881 CAS Number: 23138-50-3 EINECS: 245-446-9 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...