9,10-Dibromoanthracene 98%

Supplier: Thermo Scientific Chemicals
Warning

L12680-06 L12680-14 L12680-22
CAAAAL12680-06EA 59.25 CAD
CAAAAL12680-06 CAAAAL12680-14 CAAAAL12680-22
9,10-Dibromoanthracene 98%
9,10-Dibromoanthracene

Purity: 98% CAS No.: 523-27-3 Molecular Formula: C14H8Br2 Formula Weight: 336.04 Melting Point: 222-223° subl. Beilstein Reference: 1912109 Merck Reference: 14,3018 MDL No.: MFCD00001244

Formula: C₁₄H₈Br₂
Boiling Pt: 427 °C (1013 hPa)
Melting Pt: 222…223 °C (subl.)
Density: 1.981 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001244
CAS Number: 523-27-3
EINECS: 208-342-4
Merck Index: 14,3018

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR