2,6-Dichlorophenyl isothiocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12672-03
L12672-06
CAAAAL12672-03EA
85.95
CAD
CAAAAL12672-03
CAAAAL12672-06
2,6-Dichlorophenyl isothiocyanate 98+%
2,6-Dichlorophenyl isothiocyanate
Purity: 98+%
CAS No.: 6590-95-0
Molecular Formula: C7H3Cl2NS
Formula Weight: 204.08
Storage Temperature: 2°C to 8°C
Odor: Irritating
Boiling Point: 122-124°/8mm
Beilstein Reference: 639061
UN No.: 2923
MDL No.: MFCD00041056
Formula:
C₇H₃Cl₂NS Boiling Pt: 122…124 °C/8 mm Melting Pt: 42…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00041056 CAS Number: 6590-95-0 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...