(±)-3-(Methylamino)-1,2-propanediol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Methylamino)propane-1,2-diol, (±)-3-(Methylamino)-1,2-dihydroxy propane
L12653-14
L12653-22
CAAAAL12653-14EA
68.89
CAD
CAAAAL12653-14
CAAAAL12653-22
(±)-3-(Methylamino)-1,2-propanediol 99%
(±)-3-(Methylamino)-1,2-propanediol
Purity: 99%
CAS No.: 40137-22-2
Molecular Formula: C4H11NO2
Formula Weight: 105.14
Flash Point: 152°(305°F)
Refractive Index: 1.4745
Boiling Point: 239-241°
Beilstein Reference: 1733341
UN No.: 2735
MDL No.: MFCD00082732
Formula:
C₄H₁₁NO₂ Boiling Pt: 239…241 °C Density: 1.093 Flash Pt: 152 °C(305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00082732 CAS Number: 40137-22-2 EINECS: 254-809-0 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...