Dimethyl itaconate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester
L12652-22
L12652-36
CAAAAL12652-22EA
86.74
CAD
CAAAAL12652-22
CAAAAL12652-36
Dimethyl itaconate 97%
Dimethyl itaconate
Purity: 97%
CAS No.: 617-52-7
Molecular Formula: C7H10O4
Formula Weight: 158.16
Flash Point: 100°(212°F)
Melting Point: 33-39°
Boiling Point: 207-208°
Beilstein Reference: 386674
MDL No.: MFCD00008446
Formula:
C₇H₁₀O₄ MW: 158.15 g/mol Boiling Pt: 208 Melting Pt: 38 Density: 1.124 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008446 CAS Number: 617-52-7 EINECS: 210-519-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...