Triethyl citrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester

L12639-0B L12639-14 L12639-30
CAAAAL12639-0BEA 149.31 CAD
CAAAAL12639-0B CAAAAL12639-14 CAAAAL12639-30
Triethyl citrate 99%
Triethyl citrate

Purity: 99% CAS No.: 77-93-0 Molecular Formula: C12H20O7 Formula Weight: 276.29 Flash Point: 176°(348°F) Refractive Index: 1.4420 Melting Point: -55° Boiling Point: 126-127°/1mm Beilstein Reference: 1801199 Merck Reference: 14,2326 MDL No.: MFCD00009201

Formula: C₁₂H₂₀O₇
Boiling Pt: 126…127 °C/1 mm
Melting Pt: –55 °C
Density: 1.137
Flash Pt: 176 °C(348 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009201
CAS Number: 77-93-0
EINECS: 201-070-7
Merck Index: 14,2326

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR