Methyl-4-bromobutyrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12636-09
L12636-18
CAAAAL12636-09EA
77.58
CAD
CAAAAL12636-09
CAAAAL12636-18
Methyl-4-bromobutyrate 97%
Methyl 4-bromobutyrate
Purity: 97%
CAS No.: 4897-84-1
Molecular Formula: Br(CH2)3CO2CH3
Formula Weight: 181.04
Refractive Index: 1.4620
Boiling Point: 186-187°
MDL No.: MFCD00041482
Formula:
C₅H₉BrO₂ Boiling Pt: 186…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041482 CAS Number: 4897-84-1 EINECS: 225-523-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...