2,4,6-Trimethylphenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12611-03
L12611-06
CAAAAL12611-03EA
48.03
CAD
CAAAAL12611-03
CAAAAL12611-06
2,4,6-Trimethylphenyl isocyanate 98%
2,4,6-Trimethylphenyl isocyanate
Purity: 98%
CAS No.: 2958-62-5
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Odor: Irritating
Melting Point: 43-46°
Boiling Point: 218-220°
Beilstein Reference: 2085601
UN No.: 2811
MDL No.: MFCD00019909
Formula:
C₁₀H₁₁NO MW: 161.2 g/mol Boiling Pt: 218…220 °C Melting Pt: 43…46 °C Flash Pt: 104 °C (219 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00019909 CAS Number: 2958-62-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...