trans-2-Octene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L12606-06
L12606-14
CAAAAL12606-06EA
134.62
CAD
CAAAAL12606-06
CAAAAL12606-14
trans-2-Octene 97%
trans-2-Octene
Purity: 97%
CAS No.: 13389-42-9
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 14°(57°F)
Refractive Index: 1.4130
Melting Point: -89 to -86°
Boiling Point: 122-123°
Beilstein Reference: 1719497
UN No.: 3295
MDL No.: MFCD00009532
Formula:
C₈H₁₆ Boiling Pt: 122…123 °C Melting Pt: –89 to –86 °C Density: 0.718 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009532 CAS Number: 13389-42-9 EINECS: 236-463-2 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...